Learn More
Biomatik Corporation Rat Leukemia Inhibitory Factor, Lif Elisa Kit
Supplier:  Biomatik Corporation EKC39335
Encompass_Preferred
Rat Leukemia Inhibitory Factor, Lif Elisa Kit for detecting Leukemia Inhibitory Factor (Cholinergic Differentiation Factor) in serum, plasma, bronchoalveolar lavage fluid, tissue homogenates. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.312-20ng/ml. Sensitivity: 0.078ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P17777; Lif. Target Synonyms: LifLeukemia inhibitory factor; LIF; Cholinergic neuronal differentiation factor. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4600