Learn More
Biomatik Corporation Rat Neuroglobin, Ngb Elisa Kit
Supplier:  Biomatik Corporation EKC39472
Encompass_Preferred
Rat Neuroglobin, Ngb Elisa Kit for detecting Neuroglobin in serum, plasma, cell culture supernates, tissue homogenates. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 15.6-1000ng/ml. Sensitivity: 3.9ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q99JA8; Ngb. Target Synonyms: Ngb; Neuroglobin. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4614