Learn More
Biomatik Corporation Rat Phosphotylinosital 3 Kinase, Pi3K Elisa Kit
Supplier:  Biomatik Corporation EKC39545
Encompass_Preferred
Rat Phosphotylinosital 3 Kinase, Pi3K Elisa Kit for detecting Phosphotylinosital 3 Kinase, Pi3K in serum, plasma, tissue homogenates, cell lysates. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 15.6-1000pg/ml. Sensitivity: 3.9pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4624