Learn More
Biomatik Corporation Rat Pulmonary surfactant-associated protein A,SP-A ELISA Kit
Supplier:  Biomatik Corporation EKC39624
Encompass_Preferred
Sample Type: serum, plasma, tissue homogenates. Species Reactivity: Rattus norvegicus (Rat). Detection Range: 31.25 pg/ml - 2000 pg/ml. Sensitivity: 7.8 pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: P08427. Target Synonyms: Not Available. Restriction: For Research Use Only.
Catalog No. 50-229-4635