Learn More
Biomatik Corporation Rat Retinoic Acid Receptor Responder Protein 2 (Rarres2) Elisa Kit
Supplier:  Biomatik Corporation EKC39648
Encompass_Preferred
Rat Retinoic Acid Receptor Responder Protein 2 (Rarres2) Elisa Kit for detecting Retinoic Acid Receptor Responder (Tazarotene Induced) 2 in serum, plasma, cell culture supernates, tissue homogenates. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.9-60ng/ml. Sensitivity: 0.225ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Target Synonyms: HP10433; TIG2; chemerin. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4637