Learn More
Biomatik Corporation Rat Superoxide dismutase [Mn], mitochondrial(SOD2) ELISA kit
Supplier:  Biomatik Corporation EKC39732
Encompass_Preferred
Sample Type: serum, plasma, tissue homogenates. Species Reactivity: Rattus norvegicus (Rat). Detection Range: 1.87 pg/mL-120 pg/mL. Sensitivity: 0.47 pg/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: P07895. Target Synonyms: IPO-B, MNSOD, MVCD6, Mn-SOD, Mn superoxide dismutase|indophenoloxidase B|manganese-containing superoxide dismutase|mangano-superoxide dismutase. Restriction: For Research Use Only.
Catalog No. 50-229-4647