Learn More
Biomatik Corporation Rat Syndecan-2 (Sdc2) Elisa Kit
Supplier:  Biomatik Corporation EKC39742
Encompass_Preferred
Rat Syndecan-2 (Sdc2) Elisa Kit for detecting Syndecan 2 in serum, plasma, tissue homogenates, cell lysates. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 3.12-200ng/ml. Sensitivity: 0.78ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P34900; Sdc2. Target Synonyms: Sdc2; Hspg1; Synd2; Syndecan-2; SYND2; Fibroglycan; Heparan sulfate proteoglycan core protein; HSPG; CD antigen CD362. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4649