Learn More
Biomatik Corporation Rat thyrotropin-releasing hormone,TRH ELISA KIT
Supplier:  Biomatik Corporation EKC39778
Encompass_Preferred
Sample Type: serum, plasma, tissue homogenates. Species Reactivity: Rattus norvegicus (Rat). Detection Range: 0.5 pg/mL-100 pg/mL. Sensitivity: 0.25 pg/mL. Format: 96-Well Plate. Assay Type: Competitive. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: P01150. Target Synonyms: MGC125964, MGC125965. Restriction: For Research Use Only.
Catalog No. 50-229-4653