Learn More
Biomatik Corporation Rat Tripartite Motif-Containing Protein 72 (Trim72) Elisa Kit
Supplier:  Biomatik Corporation EKC39821
Encompass_Preferred
Rat Tripartite Motif-Containing Protein 72 (Trim72) Elisa Kit for detecting Tripartite Motif-Containing 72 in serum, plasma, tissue homogenates, cell lysates. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 18.75-1200pg/ml. Sensitivity: 4.7pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: A0JPQ4; Trim72. Target Synonyms: Trim72; Mg53; Tripartite motif-containing protein 72; Mitsugumin-53; Mg53. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4655