Learn More
Biomatik Corporation Rat α-enolase ELISA Kit
Supplier:  Biomatik Corporation EKC39892
Encompass_Preferred
Sample Type: serum, plasma, tissue homogenates. Species Reactivity: Rattus norvegicus (Rat). Detection Range: 0.625 ng/mL-40 ng/mL. Sensitivity: 0.156 ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: P04764. Target Synonyms: ENO1L1, MBP-1, MPB1, NNE, PPH, 2-phospho-D-glycerate hydro-lyase|MYC promoter-binding protein 1|alpha enolase like 1|enolase 1|non-neural enolase|phosphopyruvate hydratase|tau-crystallin |α-enolase|alpha-enolase. Restriction: For Research Use Only.
Catalog No. 50-229-4665