Learn More
Biomatik Corporation Sheep adiponectin (ADIPOQ)ELISA kit
Supplier:  Biomatik Corporation EKC39908
Encompass_Preferred
Sample Type: serum, plasma. Species Reactivity: Ovis aries (Sheep). Detection Range: 6.25 ng/mL-100 ng/mL. Sensitivity: 3.125 ng/mL. Format: 96-Well Plate. Assay Type: Competitive. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: Not Available. Target Synonyms: Not Available. Restriction: For Research Use Only.
Catalog No. 50-229-4667