Learn More
Biomatik Corporation Human Activin A, Acv-A Elisa Kit
Supplier:  Biomatik Corporation EKC40023
Encompass_Preferred
Human Activin A, Acv-A Elisa Kit for detecting Activin A, Acv-A in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 15.6-1000pg/ml. Sensitivity: 3.9pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4676