Learn More
Biomatik Corporation Mouse Insulin-like growth factor binding protein 3,IGFBP-3 ELISA KIT
Supplier:  Biomatik Corporation EKC40063
Encompass_Preferred
Sample Type: serum, plasma, tissue homogenates. Species Reactivity: Mus musculus (Mouse). Detection Range: 0.391 ng/mL-25 ng/mL. Sensitivity: 0.196 ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: P47878. Target Synonyms: tcag7.703, BP-53, IBP3, IGF-binding protein 3|acid stable subunit of the 140 K IGF complex|binding protein 29|binding protein 53|growth hormone-dependent binding protein. Restriction: For Research Use Only.
Catalog No. 50-229-4690