Learn More
Biomatik Corporation Mouse Interleukin 15,IL-15 ELISA Kit
Supplier:  Biomatik Corporation EKC40072
Encompass_Preferred
Sample Type: serum, plasma, tissue homogenates. Species Reactivity: Mus musculus (Mouse). Detection Range: 0.3125ng/ml - 20ng/ml. Sensitivity: 0.078 ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: P48346. Target Synonyms: IL-15, MGC9721. Restriction: For Research Use Only.
Catalog No. 50-229-4692