Learn More
Biomatik Corporation Human Stromal Cell Derived Factor 1Β (Sdf-1Β/Sdf1B) Elisa Kit
Supplier:  Biomatik Corporation EKC40115
Encompass_Preferred
Human Stromal Cell Derived Factor 1Β (Sdf-1Β/Sdf1B) Elisa Kit for detecting Stromal Cell Derived Factor 1Β in serum, plasma, cell culture supernates, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 125-8000pg/ml. Sensitivity: 31.25pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P48061; CXCL12. Target Synonyms: 12-O-tetradecanoylphorbol 13-acetate repressed protein 1; AI174028; C-X-C motif chemokine 12; Chemokine (C-X-C motif) ligand 12 (stromal cell-derived factor 1); Chemokine (C-X-C motif) ligand 12; Chemokine CXC motif ligand 12; cxcl12; hIRH; hSDF-1; Intercrine reduced in hepatomas; IRH; OTTHUMP00000019491; PBSF; Pre-B cell growth-stimulating factor; SCYB12; SDF 1; SDF-1; SDF-1-alpha (3-67); SDF-1a; SDF-1b; SDF1_HUMAN; SDF1A; SDF1B; Stromal cell-derived factor 1; Stromal cell-derived factor 1 delta. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4699