Learn More
Biomatik Corporation Human Vascular Endothelial Cell Growth Factor D, Vegf-D Elisa Kit
Supplier:  Biomatik Corporation EKC40130
Encompass_Preferred
Human Vascular Endothelial Cell Growth Factor D, Vegf-D Elisa Kit for detecting C-Fos Induced Growth Factor (Vascular Endothelial Growth Factor D) in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 31.25-2000pg/ml. Sensitivity: 7.81pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: O43915; VEGFD. Target Synonyms: c-fos induced growth factor; c-fos induced growth factor (vascular endothelial growth factor D); c-fos induced growth factor; c-fos-induced growth factor; FIGF; Vascular endothelial growth factor D; Vascular endothelial growth factor D precursor; Vascular endothelial growth factor D precursor; VEGF D; VEGF-D; VEGFD; VEGFD_HUMAN. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4705