Learn More
Biomatik Corporation Human Myelin Basic Protein (Mbp) Antibody Elisa Kit
Catalog No. 502294708
Encompass_Preferred
Human Myelin Basic Protein (Mbp) Antibody Elisa Kit for detecting Myelin Basic Protein(Mbp) Antibody in serum, cerebrospinal fluid (csf). Species Reactivity: Human (Homo sapiens). Detection Range: 0.625-40ng/ml. Sensitivity: 0.156ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
This item is not returnable. View return policy
Catalog No. 50-229-4708 Supplier Biomatik Corporation Supplier No. EKC40141
Add to Cart
Add to Cart
Human Myelin Basic Protein (Mbp) Antibody Elisa Kit for detecting Myelin Basic Protein(Mbp) Antibody in serum, cerebrospinal fluid (csf). Species Reactivity: Human (Homo sapiens). Detection Range: 0.625-40ng/ml. Sensitivity: 0.156ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Restrictions: For Research Use Only. Not for use in diagnostic procedures.