Learn More
Biomatik Corporation Human thyroxine,T4 ELISA Kit
Supplier:  Biomatik Corporation EKC40186
Encompass_Preferred
Sample Type: serum, plasma. Species Reactivity: Homo sapiens (Human). Detection Range: 20 ng/mL-320 ng/mL. Sensitivity: 20 ng/mL. Format: 96-Well Plate. Assay Type: Competitive. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: Not Available. Target Synonyms: Not Available. Restriction: For Research Use Only.
Catalog No. 50-229-4710