Learn More
Biomatik Corporation Rat Erythropoietin,EPO ELISA Kit
Supplier:  Biomatik Corporation EKC40292
Encompass_Preferred
Sample Type: serum, plasma, tissue homogenates. Species Reactivity: Rattus norvegicus (Rat). Detection Range: 0.156 ng/mL-10 ng/mL. Sensitivity: 0.039 ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: P29676. Target Synonyms: EP, MGC138142, MVCD2, epoetin. Restriction: For Research Use Only.
Catalog No. 50-229-4723