Learn More
Biomatik Corporation Mouse Acidic Fibroblast Growth Factor, Afgf/Fgf-1 Elisa Kit
Supplier:  Biomatik Corporation EKC40295
Encompass_Preferred
Mouse Acidic Fibroblast Growth Factor, Afgf/Fgf-1 Elisa Kit for detecting Fibroblast Growth Factor 1 (Acidic) in serum, plasma, tissue homogenates. Species Reactivity: Mouse (Mus musculus). Detection Range: 6.25-400pg/ml. Sensitivity: 1.56pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P61148; Fgf1. Target Synonyms: Fgf1; Fgf-1; Fgfa; Fibroblast growth factor 1; FGF-1; Acidic fibroblast growth factor; aFGF; Heparin-binding growth factor 1; HBGF-1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4725