Learn More
Biomatik Corporation Mouse Insulin-like growth factor binding protein 4,IGFBP-4 ELISA kit
Supplier:  Biomatik Corporation EKC40304
Encompass_Preferred
Sample Type: serum, plasma. Species Reactivity: Mus musculus (Mouse). Detection Range: 0.78 ng/mL-50 ng/mL. Sensitivity: 0.195 ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: P47879. Target Synonyms: BP-4, HT29-IGFBP, IBP4, IGFBP-4, IGF-binding protein 4|insulin-like growth factor-binding protein 4. Restriction: For Research Use Only.
Catalog No. 50-229-4727