Learn More
Biomatik Corporation Rat Urokinase-type Plasminogen Activator Receptor,PLAUR/uPAR ELISA kit
Supplier:  Biomatik Corporation EKC40311
Encompass_Preferred
Sample Type: serum, plasma, tissue homogenates. Species Reactivity: Rattus norvegicus (Rat). Detection Range: 0.156 ng/mL-10 ng/mL. Sensitivity: 0.039 ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: P49616. Target Synonyms: CD87, UPAR, URKR, monocyte activation antigen Mo3|u-plasminogen activator receptor form 2. Restriction: For Research Use Only.
Catalog No. 50-229-4729