Learn More
Biomatik Corporation Rat Tumor necrosis factor β,TNF-β ELISA KIT
Supplier:  Biomatik Corporation EKC40313
Encompass_Preferred
Sample Type: serum, plasma, tissue homogenates. Species Reactivity: Rattus norvegicus (Rat). Detection Range: 7.8 pg/mL-500 pg/mL. Sensitivity: 1.95 pg/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: Q06332. Target Synonyms: tumor necrosis factor beta, TNF-beta. Restriction: For Research Use Only.
Catalog No. 50-229-4731