Learn More
Biomatik Corporation Rat factor B,BF ELISA Kit
Supplier:  Biomatik Corporation EKC40319
Encompass_Preferred
Sample Type: serum, plasma, tissue homogenates. Species Reactivity: Rattus norvegicus (Rat). Detection Range: 1.56 ng/mL-100 ng/mL. Sensitivity: 3.9 nIU/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: Q6MG74. Target Synonyms: Not Available. Restriction: For Research Use Only.
Catalog No. 50-229-4733