Learn More
Biomatik Corporation Rat matrix metalloproteinase 10,MMP-10 ELISA kit
Supplier:  Biomatik Corporation EKC40324
Encompass_Preferred
Sample Type: serum, plasma, tissue homogenates. Species Reactivity: Rattus norvegicus (Rat). Detection Range: 80 pg/mL-2000 pg/mL. Sensitivity: 50 pg/mL. Format: 96-Well Plate. Assay Type: Competitive. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: P07152. Target Synonyms: SL-2, STMY2, matrix metalloprotease 10|matrix metalloproteinase 10|matrix metalloproteinase 10 (stromelysin 2)|stromelysin 2|transin 2. Restriction: For Research Use Only.
Catalog No. 50-229-4736