Learn More
Biomatik Corporation Rat Macrophage Colony-Stimulating Factor, M-Csf Elisa Kit
Supplier:  Biomatik Corporation EKC40328
Encompass_Preferred
Rat Macrophage Colony-Stimulating Factor, M-Csf Elisa Kit for detecting Colony Stimulating Factor 1 (Macrophage) in serum, plasma, cell culture supernates, tissue homogenates. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 15.6-1000pg/ml. Sensitivity: 3.9pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q8JZQ0; Csf1. Target Synonyms: Csf1; CsfmMacrophage colony-stimulating factor 1; CSF-1; MCSF) [Cleaved into: Processed macrophage colony-stimulating factor 1]. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4738