Learn More
Biomatik Corporation Canine Dihydrotestosterone, Dht Elisa Kit
Supplier:  Biomatik Corporation EKC40342
Encompass_Preferred
Canine Dihydrotestosterone, Dht Elisa Kit for detecting Dihydrotestosterone, Dht in serum, plasma, tissue homogenates. Species Reactivity: Dog (Canis familiaris; Canine). Detection Range: 40-8000pg/ml. Sensitivity: 20pg/ml. Format: 96-Well Plate. Assay Type: Competitive. Assay Time: 1-5h. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4742