Learn More
Biomatik Corporation Rat Thrombomodulin, Tm Elisa Kit
Supplier:  Biomatik Corporation EKC40352
Encompass_Preferred
Rat Thrombomodulin, Tm Elisa Kit for detecting Thrombomodulin, Tm in serum, plasma, tissue homogenates, cell lysates. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 1.56-100ng/ml. Sensitivity: 0.39ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4744