Learn More
Biomatik Corporation Dog Interleukin-8, Il-8 Elisa Kit
Supplier:  Biomatik Corporation EKC40374
Encompass_Preferred
Dog Interleukin-8, Il-8 Elisa Kit for detecting Interleukin-8, Il-8 in serum, plasma, cell culture supernates, tissue homogenates. Species Reactivity: Dog (Canis familiaris; Canine). Detection Range: 0.312-20ng/ml. Sensitivity: 0.146ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P41324; CXCL8. Target Synonyms: CXCL8; IL8; Interleukin-8; IL-8; C-X-C motif chemokine 8; Chemokine; C-X-C motif) ligand 8. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4747