Learn More
Biomatik Corporation Human B-Cell Leukemia/Lymphoma 2, Bcl-2 Elisa Kit
Supplier:  Biomatik Corporation EKC40526
Encompass_Preferred
Human B-Cell Leukemia/Lymphoma 2, Bcl-2 Elisa Kit for detecting B-Cell Cll/Lymphoma 2 in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 0.9-60ng/ml. Sensitivity: 0.225ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P10415; BCL2. Target Synonyms: BCL2; Apoptosis regulator Bcl-2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4763