Learn More
Biomatik Corporation Human Cxc-Chemokine Ligand 16, Cxcl16 Elisa Kit
Supplier:  Biomatik Corporation EKC40532
Encompass_Preferred
Human Cxc-Chemokine Ligand 16, Cxcl16 Elisa Kit for detecting Chemokine (C-X-C Motif) Ligand 16 in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 46.875-3000pg/ml. Sensitivity: 3.603pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q9H2A7; CXCL16. Target Synonyms: C-X-C motif chemokine 16; Chemokine (C X C motif) ligand 16; Chemokine; CXC motif; ligand 16; CXC chemokine ligand 16; Cxcl16; CXCLG16; CXL16_HUMAN; Scavenger receptor for phosphatidylserine and oxidized low density lipoprotein; SCYB16; Small inducible cytokine B16 precursor; Small-inducible cytokine B16; SR-PSOX; SRPSOX; Transmembrane chemokine CXCL16; UNQ2759/PRO6714. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4766