Learn More
Biomatik Corporation Human Soluble Cluster Of Differentiation 38, Scd38 Elisa Kit
Supplier:  Biomatik Corporation EKC40704
Encompass_Preferred
Human Soluble Cluster Of Differentiation 38, Scd38 Elisa Kit for detecting Cd38 Molecule in serum, plasma, tissue homogenates, cell lysates. Species Reactivity: Human (Homo sapiens). Detection Range: 0.9-60ng/ml. Sensitivity: 0.225ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P28907; CD38. Target Synonyms: CD38; ADP-ribosyl cyclase/cyclic ADP-ribose hydrolase 1; 2'-phospho-ADP-ribosyl cyclase; 2'-phospho-ADP-ribosyl cyclase/2'-phospho-cyclic-ADP-ribose transferase; 2'-phospho-cyclic-ADP-ribose transferase; ADP-ribosyl cyclase 1; ADPRC 1; Cyclic ADP-ribose hydrolase 1; cADPr hydrolase 1; T10; CD antigen CD38. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4784