Learn More
Biomatik Corporation Human Interleukin 1 Receptor Antagonist, Il-1Ra Elisa Kit
Supplier:  Biomatik Corporation EKC40762
Encompass_Preferred
Human Interleukin 1 Receptor Antagonist, Il-1Ra Elisa Kit for detecting Interleukin 1 Receptor Antagonist in serum, plasma, cell culture supernates, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 15.6-1000pg/ml. Sensitivity: 3.9pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P18510; IL1RN. Target Synonyms: DIRA; F630041P17Rik; ICIL 1RA; ICIL-1RA; ICIL1RA; IL-1ra; IL-1ra3; IL-1RN; IL1 inhibitor; IL1F3; IL1RA; IL1RA_HUMAN; IL1RN (IL1F3); IL1RN; Interleukin 1 receptor antagonist; Interleukin-1 receptor antagonist protein; Intracellular IL 1 receptor antagonist type II; Intracellular interleukin 1 receptor antagonist (icIL 1ra); IRAP; MGC10430; MVCD4; Type II interleukin 1 receptor antagonist. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4793