Learn More
Biomatik Corporation Human Atp-Binding Cassette Transporter G2, Abcg2 Elisa Kit
Supplier:  Biomatik Corporation EKC40789
Encompass_Preferred
Human Atp-Binding Cassette Transporter G2, Abcg2 Elisa Kit for detecting Atp-Binding Cassette, Sub-Family G (White), Member 2 in serum, plasma, cell lysates, cell culture supernates. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.039ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q9UNQ0; ABCG2. Target Synonyms: ABCG2; ABCP; BCRP; BCRP1; MXR; Broad substrate specificity ATP-binding cassette transporter ABCG2; ATP-binding cassette sub-family G member 2; Breast cancer resistance protein; CDw338; Mitoxantrone resistance-associated protein; Placenta-specific ATP-binding cassette transporter; Urate exporter; CD antigen CD338. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4795