Learn More
Biomatik Corporation Human Lectin-Galactose Binding-Soluble 3, Lgals3 Elisa Kit
Supplier:  Biomatik Corporation EKC40817
Encompass_Preferred
Human Lectin-Galactose Binding-Soluble 3, Lgals3 Elisa Kit for detecting Lectin, Galactoside-Binding, Soluble, 3 in serum, plasma, cell culture supernates, tissue homogenates, saliva. Species Reactivity: Human (Homo sapiens). Detection Range: 0.625-40ng/ml. Sensitivity: 0.156ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P17931; LGALS3. Target Synonyms: 35 kDa lectin; Carbohydrate binding protein 35; Carbohydrate-binding protein 35; CBP 35; CBP35; Gal-3; GAL3; Galactose-specific lectin 3; Galactoside-binding protein; GALBP; Galectin 3 internal gene; included; Galectin-3; Galectin3; GALIG; GBP; IgE binding protein; IgE-binding protein; L 31; L 34; L-31; L-34 galactoside-binding lectin; L31; Laminin-binding protein; Lectin L-29; Lectin; galactose binding; soluble 3; LGALS2; LGALS3; MAC 2 antigen; Mac-2; Mac-2 antigen; MAC2; Macrophage galactose-specific lectin; MGC105387. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4799