Learn More
Biomatik Corporation Human A Disintegrin And Metalloprotease 9, Adam9 Elisa Kit
Supplier:  Biomatik Corporation EKC40829
Encompass_Preferred
Human A Disintegrin And Metalloprotease 9, Adam9 Elisa Kit for detecting Adam Metallopeptidase Domain 9 (Meltrin Gamma) in serum, plasma, tissue homogenates, cell culture supernates, ascitic fluid. Species Reactivity: Human (Homo sapiens). Detection Range: 62.5-4000pg/ml. Sensitivity: 15.6pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q13443; ADAM9. Target Synonyms: ADAM 9; ADAM metallopeptidase domain 9; Adam9; ADAM9_HUMAN; Cellular disintegrin-related protein; Cone rod dystrophy 9; CORD9; Disintegrin and metalloproteinase domain-containing protein 9; MCMP; MDC9; Meltrin-gamma; Metalloprotease/disintegrin/cysteine-rich protein 9; Mltng; Myeloma cell metalloproteinase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4803