Learn More
Biomatik Corporation Rat Collagen alpha-1(I) chain (COL1A1) ELISA kit
Supplier:  Biomatik Corporation EKC40873
Encompass_Preferred
Sample Type: serum, plasma, tissue homogenates. Species Reactivity: Rattus norvegicus (Rat). Detection Range: 0.156 ng/mL-10 ng/mL. Sensitivity: 0.039 ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: P02456. Target Synonyms: OI4, alpha 1 type I collagen|collagen alpha 1 chain type I|collagen of skin, tendon and bone, alpha-1 chain|pro-alpha-1 collagen type 1,_x000D_,Collagen alpha-1(1) chain (COL1A1),_x000D_,Collagen alpha-1(I) chain (COL1A1). Restriction: For Research Use Only.
Catalog No. 50-229-4807