Learn More
Biomatik Corporation Bovine Beta-Endorphin, Β-Ep Elisa Kit
Supplier:  Biomatik Corporation EKC40924
Encompass_Preferred
Bovine Beta-Endorphin, Β-Ep Elisa Kit for detecting Beta-Endorphin, Β-Ep in serum, plasma, tissue homogenates. Species Reactivity: Bovine (Bos taurus). Detection Range: 46.88-3000pg/ml. Sensitivity: 11.72pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Target Synonyms: beta-Endorphin; beta-EP. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4810