Learn More
Biomatik Corporation Human Nidogen 1 (Nid1) Elisa Kit
Supplier:  Biomatik Corporation EKC40970
Encompass_Preferred
Human Nidogen 1 (Nid1) Elisa Kit for detecting Nidogen 1 in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 0.39-25ng/ml. Sensitivity: 0.09ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P14543; NID1. Target Synonyms: Enactin; Ent; Entactin 1; Entactin; NID 1; NID; NID-1; Nid1; NID1_HUMAN; Nidogen (enactin); Nidogen 1; Nidogen; Nidogen-1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4813