Learn More
Biomatik Corporation Human Galectin 7 (Gal-7) Elisa Kit
Supplier:  Biomatik Corporation EKC41010
Encompass_Preferred
Human Galectin 7 (Gal-7) Elisa Kit for detecting Galectin 7(Gal-7) in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 78-5000pg/ml. Sensitivity: 19.5pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P47929; LGALS7. Target Synonyms: Gal-7; GAL7; Galectin 7B; Galectin-7; HKL-14; Human keratinocyte lectin 14; Keratinocyte lectin 14; Lectin galactoside binding soluble 7; Lectin; galactoside binding; soluble; 7B; LEG7_HUMAN; LGALS7; LGALS7A; LGALS7B; P53 induced protein 1; p53-induced gene 1 protein; Pi7; PIG1; TP53I1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4819