Learn More
Biomatik Corporation Dog Matrix Metalloproteinase 3, Mmp-3 Elisa Kit
Supplier:  Biomatik Corporation EKC41036
Encompass_Preferred
Dog Matrix Metalloproteinase 3, Mmp-3 Elisa Kit for detecting Matrix Metallopeptidase 3 (Stromelysin 1, Progelatinase) in serum, plasma, cell culture supernates, urine, tissue homogenates. Species Reactivity: Dog (Canis familiaris; Canine). Detection Range: 1.56-100ng/ml. Sensitivity: 0.39ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q6Y4Q5; MMP3. Target Synonyms: MMP3Stromelysin-1; SL-1; EC 3.4.24.17; Matrix metalloproteinase-3; MMP-3. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4826