Learn More
Biomatik Corporation Human Major Histocompatibility Complex Class I Chain-Related Gene A (Mica) Elisa Kit
Supplier:  Biomatik Corporation EKC41061
Encompass_Preferred
Human Major Histocompatibility Complex Class I Chain-Related Gene A (Mica) Elisa Kit for detecting Mhc Class I Polypeptide-Related Sequence A in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 1.56-100ng/ml. Sensitivity: 0.39ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q29983; MICA. Target Synonyms: MHC class I chain-related protein A; MHC class I chain-related protein B; MHC class I polypeptide related sequence A; MHC class I polypeptide related sequence B; MHC class I polypeptide-related sequence A; MIC-A; micA; MICA_HUMAN; MICB. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4830