Learn More
Biomatik Corporation Rat Glucose Transporter 4 (Glut4) Elisa Kit
Supplier:  Biomatik Corporation EKC41078
Encompass_Preferred
Rat Glucose Transporter 4 (Glut4) Elisa Kit for detecting Solute Carrier Family 2 (Facilitated Glucose Transporter), Member 4 in serum, plasma, tissue homogenates, cell lysates. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.039ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P19357; Slc2a4. Target Synonyms: Slc2a4; Glut4Solute carrier family 2; facilitated glucose transporter member 4; Glucose transporter type 4; insulin-responsive; GLUT-4. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4834