Learn More
Biomatik Corporation Human Selenium Binding Protein 1 (Selenbp1) Elisa Kit
Supplier:  Biomatik Corporation EKC41085
Encompass_Preferred
Human Selenium Binding Protein 1 (Selenbp1) Elisa Kit for detecting Selenium Binding Protein 1 in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 125-8000pg/ml. Sensitivity: 31.25pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q13228; SELENBP1. Target Synonyms: 56 kDa selenium binding protein; 56 kDa selenium-binding protein; hSBP; LPSB; SBP; SBP1; SBP1_HUMAN; SBP56; Selenbp1; Selenbp2; Selenium Binding Protein 1; Selenium binding protein 2; Selenium binding protein1; Selenium-binding protein 1; SELNBP1; SP56. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4836