Learn More
Biomatik Corporation Human Thymidine Kinase 1, Soluble (Tk1) Elisa Kit
Supplier:  Biomatik Corporation EKC41132
Encompass_Preferred
Human Thymidine Kinase 1, Soluble (Tk1) Elisa Kit for detecting Thymidine Kinase 1, Soluble in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 62.5-4000pg/ml. Sensitivity: 15.6pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P04183; TK1. Target Synonyms: cytosolic; KITH_HUMAN; Thymidine kinase 1; Thymidine kinase 1 soluble; Thymidine kinase 1 soluble isoform; Thymidine kinase; Thymidine kinase cytosolic; TK 1; TK 2; TK1; Tk1a; Tk1b; TK2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4842