Learn More
Biomatik Corporation Canine Cardiac Isoform Of Tropnin T, Ctnt Elisa Kit
Supplier:  Biomatik Corporation EKC41146
Encompass_Preferred
Canine Cardiac Isoform Of Tropnin T, Ctnt Elisa Kit for detecting Cardiac Isoform Of Tropnin T, Ctnt in serum, plasma, tissue homogenates. Species Reactivity: Dog (Canis familiaris; Canine). Detection Range: 15.6-1000pg/ml. Sensitivity: 3.9pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: J9P6P7. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4844