Learn More
Biomatik Corporation Human Soluble Cluster Of Differentiation 80 (Scd80) Elisa Kit
Supplier:  Biomatik Corporation EKC41174
Encompass_Preferred
Human Soluble Cluster Of Differentiation 80 (Scd80) Elisa Kit for detecting Cd80 Molecule in serum, plasma, cell culture supernates. Species Reactivity: Human (Homo sapiens). Detection Range: 125-8000pg/ml. Sensitivity: 31.25pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P33681; CD80. Target Synonyms: CD80; CD28LG; CD28LG1; LAB7; T-lymphocyte activation antigen CD80; Activation B7-1 antigen; BB1; CTLA-4 counter-receptor B7.1; B7; CD antigen CD80. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4848