Learn More
Biomatik Corporation Mouse NAD-dependent deacetylase sirtuin-1(Sirt1)ELISA Kit
Supplier:  Biomatik Corporation EKC41206
Encompass_Preferred
Sample Type: serum, plasma, tissue homogenates. Species Reactivity: Mus musculus (Mouse). Detection Range: 0.156 ng/mL-10 ng/mL. Sensitivity: 0.039 ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: Q923E4. Target Synonyms: RP11-57G10.3, SIR2L1, SIR2alpha|sir2-like 1|sirtuin 1|sirtuin type 1. Restriction: For Research Use Only.
Catalog No. 50-229-4852