Learn More
Biomatik Corporation Human Endothelin-2 (Edn2) Elisa Kit
Supplier:  Biomatik Corporation EKC41230
Encompass_Preferred
Human Endothelin-2 (Edn2) Elisa Kit for detecting Endothelin 2 in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 4.688-300pg/ml. Sensitivity: 1.172pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P20800; EDN2. Target Synonyms: EDN2; EDN2_HUMAN; Endothelin 2; Endothelin-2; ET 2; ET-2; ET2; PPET2; Preproendothelin 2; Preproendothelin-2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4854