Learn More
Biomatik Corporation Human A Disintegrin And Metalloproteinase With Thrombospondin Motifs 9 (Adamts9) Elisa Kit
Supplier:  Biomatik Corporation EKC41340
Encompass_Preferred
Human A Disintegrin And Metalloproteinase With Thrombospondin Motifs 9 (Adamts9) Elisa Kit for detecting Adam Metallopeptidase With Thrombospondin Type 1 Motif, 9 in serum, plasma, cell culture supernates, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 0.78-50ng/ml. Sensitivity: 0.195ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q9P2N4; ADAMTS9. Target Synonyms: ADAMTS9; KIAA1312; A disintegrin and metalloproteinase with thrombospondin motifs 9; ADAM-TS 9; ADAM-TS9; ADAMTS-9; EC 3.4.24. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4863